Assay Details
Amplicon Length: | 79 |
Chromosome Location: | Chr. 8 - 128150518 - 128150518 [+] on 128150518 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1208 | MIMAT0005873 |
Stem-loop Details
Human
