Assay Details
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UAGGUAGUUUCCUGUUGUUGGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-196b-5p | MIMAT0001080 |
Mouse | mmu-miR-196b-5p | MIMAT0001081 |
Rat | rno-miR-196b-5p | MIMAT0001082 |
Callithrix jacchus | cja-miR-196b | MIMAT0039444 |
Cricetulus griseus | cgr-miR-196b | MIMAT0023833 |
Gorilla gorilla | ggo-miR-196b | MIMAT0049110 |
View More Rows | ||
Horse | eca-miR-196b | MIMAT0012939 |
Microcebus murinus | mmr-miR-196b | MIMAT0049222 |
Monodelphis domestica | mdo-miR-196a-5p | MIMAT0028494 |
Nomascus leucogenys | nle-miR-196b | MIMAT0049406 |
Otolemur garnettii | oga-miR-196b | MIMAT0049655 |
Pan troglodytes | ptr-miR-196b | MIMAT0008061 |
Pig | ssc-miR-196b-5p | MIMAT0013923 |
Pongo pygmaeus | ppy-miR-196b | MIMAT0015791 |
Pteropus alecto | pal-miR-196b-5p | MIMAT0040009 |
Rhesus monkey | mml-miR-196b-5p | MIMAT0006229 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Cricetulus griseus
Gorilla gorilla
Horse
Microcebus murinus
Monodelphis domestica
Nomascus leucogenys
Otolemur garnettii
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-196b (v17)
Mouse: mmu-miR-196b (v17)
Rat: rno-miR-196b (v18)
Macaca mulatta: mml-miR-196b (v19)
Staphylococcus schleiferi: ssc-miR-196b (v20)
Mouse: mmu-miR-196b (v17)
Rat: rno-miR-196b (v18)
Macaca mulatta: mml-miR-196b (v19)
Staphylococcus schleiferi: ssc-miR-196b (v20)
