Assay Details
Amplicon Length: | 97 |
Chromosome Location: | Chr. X - 74218869 - 74218869 [+] on 74218869 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-374b-3p | MIMAT0004956 |
| Human | hsa-miR-374b-5p | MIMAT0004955 |
| Human | hsa-miR-374c-5p | MIMAT0018443 |
| Human | hsa-miR-374c-3p | MIMAT0022735 |
Stem-loop Details
Human

Genomic Map