Assay Details
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | AUAUAAUACAACCUGCUAAGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-374b-5p | MIMAT0004955 | |
| Mouse | mmu-miR-374b-5p | MIMAT0003727 | |
| Rat | rno-miR-374-5p | MIMAT0003208 | |
| Bovine | bta-miR-374b | MIMAT0009302 | |
| Cricetulus griseus | cgr-miR-374-5p | MIMAT0023939 | |
| Dog | cfa-miR-374b | MIMAT0006754 | |
| View More Rows | |||
| Goat | chi-miR-374b-5p | MIMAT0036182 | |
| Gorilla gorilla | ggo-miR-374b | MIMAT0024126 | |
| Horse | eca-miR-374b | MIMAT0013214 | |
| Pan troglodytes | ptr-miR-374b | MIMAT0008124 | |
| Pig | ssc-miR-374b-5p | MIMAT0013915 | |
| Pteropus alecto | pal-miR-374b-5p | MIMAT0039989 | |
| Rabbit | ocu-miR-374b-5p | MIMAT0048408 | |
| Rhesus monkey | mml-miR-374b-5p | MIMAT0006301 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Dog
Goat
Gorilla gorilla
Horse
Pan troglodytes
Pig
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool B v3.0Megaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-374b (v17)
Mouse: mmu-miR-374-5p (v18)
Mouse: mmu-miR-374 (v17)
Rat: rno-miR-374 (v18)
Macaca mulatta: mml-miR-374b (v19)
Staphylococcus schleiferi: ssc-miR-374b (v15)
Mouse: mmu-miR-374-5p (v18)
Mouse: mmu-miR-374 (v17)
Rat: rno-miR-374 (v18)
Macaca mulatta: mml-miR-374b (v19)
Staphylococcus schleiferi: ssc-miR-374b (v15)

Genomic Map