Assay Details
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | AUAUAAUACAACCUGCUAAGUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-374b-5p | MIMAT0004955 |
Mouse | mmu-miR-374b-5p | MIMAT0003727 |
Rat | rno-miR-374-5p | MIMAT0003208 |
Bovine | bta-miR-374b | MIMAT0009302 |
Cricetulus griseus | cgr-miR-374-5p | MIMAT0023939 |
Dog | cfa-miR-374b | MIMAT0006754 |
View More Rows | ||
Goat | chi-miR-374b-5p | MIMAT0036182 |
Gorilla gorilla | ggo-miR-374b | MIMAT0024126 |
Horse | eca-miR-374b | MIMAT0013214 |
Pan troglodytes | ptr-miR-374b | MIMAT0008124 |
Pig | ssc-miR-374b-5p | MIMAT0013915 |
Pteropus alecto | pal-miR-374b-5p | MIMAT0039989 |
Rabbit | ocu-miR-374b-5p | MIMAT0048408 |
Rhesus monkey | mml-miR-374b-5p | MIMAT0006301 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Dog
Goat
Gorilla gorilla
Horse
Pan troglodytes
Pig
Pteropus alecto
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-374b (v17)
Mouse: mmu-miR-374-5p (v18)
Mouse: mmu-miR-374 (v17)
Rat: rno-miR-374 (v18)
Macaca mulatta: mml-miR-374b (v19)
Staphylococcus schleiferi: ssc-miR-374b (v15)
Mouse: mmu-miR-374-5p (v18)
Mouse: mmu-miR-374 (v17)
Rat: rno-miR-374 (v18)
Macaca mulatta: mml-miR-374b (v19)
Staphylococcus schleiferi: ssc-miR-374b (v15)
