Assay Details
Amplicon Length: | 137 |
Chromosome Location: | Chr. 19 - 53753760 - 53753760 [+] on 53753760 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-527 | MIMAT0002862 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 137 |
Chromosome Location: | Chr. 19 - 53753760 - 53753760 [+] on 53753760 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-527 | MIMAT0002862 |