Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUGCAAAGGGAAGCCCUUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518a-5p | MIMAT0005457 |
| Human | hsa-miR-527 | MIMAT0002862 |
| Gorilla gorilla | ggo-miR-1283 | MIMAT0036431 |
| Pan troglodytes | ptr-miR-1283b | MIMAT0036458 |
| Pan troglodytes | ptr-miR-518g-5p | MIMAT0036454 |
| Pan troglodytes | ptr-miR-527 | MIMAT0008212 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-518a-5p | MIMAT0015951 |
| Pongo pygmaeus | ppy-miR-527 | MIMAT0015986 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus

Genomic Map