Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUGCAAAGGGAAGCCCUUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518a-5p | MIMAT0005457 |
| Human | hsa-miR-527 | MIMAT0002862 |
| Gorilla gorilla | ggo-miR-1283 | MIMAT0036431 |
| Pan troglodytes | ptr-miR-1283b | MIMAT0036458 |
| Pan troglodytes | ptr-miR-518g-5p | MIMAT0036454 |
| Pan troglodytes | ptr-miR-527 | MIMAT0008212 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-518a-5p | MIMAT0015951 |
| Pongo pygmaeus | ppy-miR-527 | MIMAT0015986 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-518a-1 |
| Stem-loop Accession # | MI0003170 |
| Stem-loop Sequence | UCUCAAGCUGUGACUGCAAAGGGAAGCCCUUUCUGUUGUCUGAAAGAAGAGAAAGCGCUUCCCUUUGCUGGAUUACGGUUUGAGA |
| Chromosome Location | Chr. 19 - 53731006 - 53731090 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005457
miRBase ID: hsa-miR-518a-5p Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: Chr. 19 - 53739333 - 53739419 [+] on Build GRCh38 |
| Stem-loop ID | hsa-mir-518a-2 |
| Stem-loop Accession # | MI0003173 |
| Stem-loop Sequence | UCUCAAGCUGUGGGUCUGCAAAGGGAAGCCCUUUCUGUUGUCUAAAAGAAGAGAAAGCGCUUCCCUUUGCUGGAUUACGGUUUGAGA |
| Chromosome Location | Chr. 19 - 53739333 - 53739419 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005457
miRBase ID: hsa-miR-518a-5p Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: Chr. 19 - 53739333 - 53739419 [+] on Build GRCh38 |
| Stem-loop ID | hsa-mir-527 |
| Stem-loop Accession # | MI0003179 |
| Stem-loop Sequence | UCUCAAGCUGUGACUGCAAAGGGAAGCCCUUUCUGUUGUCUAAAAGAAAAGAAAGUGCUUCCCUUUGGUGAAUUACGGUUUGAGA |
| Chromosome Location | Chr. 19 - 53754018 - 53754102 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002862
miRBase ID: hsa-miR-527 Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: Chr. 19 - 53754018 - 53754102 [+] on Build GRCh38 |
| Stem-loop ID | ggo-mir-1283-1 |
| Stem-loop Accession # | MI0031084 |
| Stem-loop Sequence | UGCUGGAGCAAGAAGAUCUCAGGCUGUGACUCUGCAAAGGGAAGCCCUUUCUGUUGUCAGAAAGAAAAGAAAGCGCUUCCCUUUGCUGGGUUACGGUUUGAGAAAGCGACGUUGAAGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036431
miRBase ID: ggo-miR-1283 Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
| Stem-loop ID | ggo-mir-1283-2 |
| Stem-loop Accession # | MI0031091 |
| Stem-loop Sequence | GGAAGAUCUCAAGCUGUGACUCUGCAAAGGGAAGCCCUUUCUGCUGUCUAAAAGAAAAGAAAGUGCUUCCCUUUGGUGAAUUACGGUUUGAGAAAAGCAGCGUUGAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036431
miRBase ID: ggo-miR-1283 Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
| Stem-loop ID | ggo-mir-1283-3 |
| Stem-loop Accession # | MI0031093 |
| Stem-loop Sequence | CUGGAGCAAGAAGAUCUCAUGCUGUGAGUCUGCAAAGGGAAGCCCUUUCUGUUGUCUGAAAGAAAAGAAAUCGCUUCCCUUUGGAGUGUUACGGUUUGAGAAAAGCAGCGUUGAAGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036431
miRBase ID: ggo-miR-1283 Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1283b |
| Stem-loop Accession # | MI0031111 |
| Stem-loop Sequence | CUGGAGCAAGAAGAUCUCAGGCUGUGAGUCUGCAAAGGGAAGCCCUUUCUGUUGUCUGAAAGAAAAGAAAUCGCUUCCCUUUGGAGUGUUACGGUUUGAGAAAAGCAGCGUUGAAGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036458
miRBase ID: ptr-miR-1283b Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
| Stem-loop ID | ptr-mir-518g |
| Stem-loop Accession # | MI0031109 |
| Stem-loop Sequence | AAGAAGAUCUCGUGCUGUGACUCUGCAAAGGGAAGCCCUUUCUGUUGUCUAAAAGAAUAGAAAGCGCUUCCCUUUGGAGUGUUACGGUUUGAGAAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036454
miRBase ID: ptr-miR-518g-5p Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
| Stem-loop ID | ptr-mir-527 |
| Stem-loop Accession # | MI0008740 |
| Stem-loop Sequence | CUCAGGCUGUGACUCUGCAAAGGGAAGCCCUUUCUGUUGUCUAAAAGAAAAGAAAGUGCUUCCCUUUGGUGAAUUACGGUUUGAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008212
miRBase ID: ptr-miR-527 Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
| Stem-loop ID | ppy-mir-518a-1 |
| Stem-loop Accession # | MI0015006 |
| Stem-loop Sequence | UCUCAUGCUGUGACUCUGCAAAGGGAAGCCCUUUCUGUUGUCUAAAAGAAAAGAAAGCGCUUCCCUUUGGAGUGUUACGGUUUGAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015951
miRBase ID: ppy-miR-518a-5p Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
| Stem-loop ID | ppy-mir-518a-2 |
| Stem-loop Accession # | MI0031108 |
| Stem-loop Sequence | CUGGAGCAAGAAGAUCUCAUGCUGUGACUCUGCAAAGGGAAGCCCUUUCUGCUGUCUAAAAGAAAAGAAAGCGCUUCCCUUUGGAGUGUUACGGUUUGAGAAAAGCAGCGUUGAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015951
miRBase ID: ppy-miR-518a-5p Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
| Stem-loop ID | ppy-mir-527 |
| Stem-loop Accession # | MI0015036 |
| Stem-loop Sequence | UCUCAUGCUGUGACUCUGCAAAGGGAAGCCCUUUCUGUUGUCUAAAAGAAAAGAAAGUGCUUCCCUUUGGUAGGUUACGGUUUGAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015986
miRBase ID: ppy-miR-527 Mature miRNA Sequence: CUGCAAAGGGAAGCCCUUUC Chromosome Location: NA |
More Information
Related Products

Genomic Map