Assay Details
Gene Family ID: | MIPF0000318, mir-488 |
Mature miRNA Sequence: | CCCAGAUAAUGGCACUCUCAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-488-5p | MIMAT0002804 |
| Rat | rno-miR-488-5p | MIMAT0017320 |
| Dog | cfa-miR-488 | MIMAT0009903 |
| Gorilla gorilla | ggo-miR-488 | MIMAT0024230 |
Stem-loop Details
Human
Rat
Dog
Gorilla gorilla

Genomic Map