Assay Details
Amplicon Length: | 79 |
Chromosome Location: | Chr. 7 - 93484236 - 93484236 [+] on 93484236 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-489-3p | MIMAT0002805 |
| Human | hsa-miR-489-5p | MIMAT0026605 |
Stem-loop Details
Human

Genomic Map