Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | GUGACAUCACAUAUACGGCAGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-489-3p | MIMAT0002805 |
Callithrix jacchus | cja-miR-489 | MIMAT0039613 |
Pan troglodytes | ptr-miR-489 | MIMAT0008164 |
Pongo pygmaeus | ppy-miR-489 | MIMAT0015908 |
Rhesus monkey | mml-miR-489-3p | MIMAT0006349 |
Stem-loop Details
Human
Callithrix jacchus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-489 (v19)
Macaca mulatta: mml-miR-489 (v19)
Macaca mulatta: mml-miR-489 (v19)
