Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | GUGACAUCACAUAUACGGCAGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-489-3p | MIMAT0002805 |
| Callithrix jacchus | cja-miR-489 | MIMAT0039613 |
| Pan troglodytes | ptr-miR-489 | MIMAT0008164 |
| Pongo pygmaeus | ppy-miR-489 | MIMAT0015908 |
| Rhesus monkey | mml-miR-489-3p | MIMAT0006349 |
Stem-loop Details
Human
Callithrix jacchus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-489 (v19)
Macaca mulatta: mml-miR-489 (v19)
Macaca mulatta: mml-miR-489 (v19)

Genomic Map