Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 9 - 28888679 - 28888679 [-] on 28888679 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-873-3p | MIMAT0022717 |
| Human | hsa-miR-873-5p | MIMAT0004953 |
Stem-loop Details
Human

Genomic Map