Assay Details
Gene Family ID: | MIPF0000390, mir-873 |
Mature miRNA Sequence: | GCAGGAACUUGUGAGUCUCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-873-5p | MIMAT0004953 |
| Mouse | mmu-miR-873a-5p | MIMAT0004936 |
| Rat | rno-miR-873-5p | MIMAT0005339 |
| Bovine | bta-miR-873 | MIMAT0009377 |
| Callithrix jacchus | cja-miR-873 | MIMAT0049330 |
| Dasypus novemcinctus | dno-miR-873-5p | MIMAT0048016 |
| View More Rows | ||
| Horse | eca-miR-873 | MIMAT0013120 |
| Pan troglodytes | ptr-miR-873 | MIMAT0008324 |
| Pongo pygmaeus | ppy-miR-873 | MIMAT0016111 |
| Pteropus alecto | pal-miR-873-5p | MIMAT0040033 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto

Genomic Map