Assay Details
Gene Family ID: | MIPF0000390, mir-873 |
Mature miRNA Sequence: | GCAGGAACUUGUGAGUCUCCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-873-5p | MIMAT0004953 | |
| Mouse | mmu-miR-873a-5p | MIMAT0004936 | |
| Rat | rno-miR-873-5p | MIMAT0005339 | |
| Bovine | bta-miR-873 | MIMAT0009377 | |
| Callithrix jacchus | cja-miR-873 | MIMAT0049330 | |
| Dasypus novemcinctus | dno-miR-873-5p | MIMAT0048016 | |
| View More Rows | |||
| Horse | eca-miR-873 | MIMAT0013120 | |
| Pan troglodytes | ptr-miR-873 | MIMAT0008324 | |
| Pongo pygmaeus | ppy-miR-873 | MIMAT0016111 | |
| Pteropus alecto | pal-miR-873-5p | MIMAT0040033 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-873 (v17)
Mouse: mmu-miR-873 (v17)
Mouse: mmu-miR-873-5p (v18)
Rat: rno-miR-873 (v18)
Mouse: mmu-miR-873 (v17)
Mouse: mmu-miR-873-5p (v18)
Rat: rno-miR-873 (v18)

Genomic Map