Assay Details
Amplicon Length: | 115 |
Chromosome Location: | Chr. 9 - 94176592 - 94176592 [+] on 94176592 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-let-7f-5p | MIMAT0000067 |
Human | hsa-let-7f-1-3p | MIMAT0004486 |
Stem-loop Details
Human
