Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAAUCUAUUGCCUUCCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-let-7f-1-3p | MIMAT0004486 |
Mouse | mmu-let-7f-1-3p | MIMAT0004623 |
Chicken | gga-let-7f-3p | MIMAT0026528 |
Columba livia | cli-let-7f-3p | MIMAT0038386 |
Dasypus novemcinctus | dno-let-7f-3p | MIMAT0047483 |
Goat | chi-let-7f-3p | MIMAT0035891 |
View More Rows | ||
Guinea pig | cpo-let-7f-3p | MIMAT0046829 |
Monodelphis domestica | mdo-let-7f-1-3p | MIMAT0026702 |
Ophiophagus hannah | oha-let-7f-2-3p | MIMAT0036638 |
Pteropus alecto | pal-let-7f-3p | MIMAT0039771 |
Rabbit | ocu-let-7f-3p | MIMAT0048085 |
Rhesus monkey | mml-let-7f-3p | MIMAT0026793 |
Salmo salar | ssa-let-7f-3p | MIMAT0032269 |
Stem-loop Details
Human
Mouse
Chicken
Columba livia
Dasypus novemcinctus
Goat
Guinea pig
Monodelphis domestica
Ophiophagus hannah
Pteropus alecto
Rabbit
Rhesus monkey
Salmo salar

More Information
Related Products
miRBase Alias:
Human: hsa-let-7f-1* (v17)
Mouse: mmu-let-7f* (v15)
Mouse: mmu-let-7f-1* (v17)
Mouse: mmu-let-7f* (v15)
Mouse: mmu-let-7f-1* (v17)
