Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAAUCUAUUGCCUUCCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7f-1-3p | MIMAT0004486 |
| Mouse | mmu-let-7f-1-3p | MIMAT0004623 |
| Chicken | gga-let-7f-3p | MIMAT0026528 |
| Columba livia | cli-let-7f-3p | MIMAT0038386 |
| Dasypus novemcinctus | dno-let-7f-3p | MIMAT0047483 |
| Goat | chi-let-7f-3p | MIMAT0035891 |
| View More Rows | ||
| Guinea pig | cpo-let-7f-3p | MIMAT0046829 |
| Monodelphis domestica | mdo-let-7f-1-3p | MIMAT0026702 |
| Ophiophagus hannah | oha-let-7f-2-3p | MIMAT0036638 |
| Pteropus alecto | pal-let-7f-3p | MIMAT0039771 |
| Rabbit | ocu-let-7f-3p | MIMAT0048085 |
| Rhesus monkey | mml-let-7f-3p | MIMAT0026793 |
| Salmo salar | ssa-let-7f-3p | MIMAT0032269 |
Stem-loop Details
Human
Mouse
Chicken
Columba livia
Dasypus novemcinctus
Goat
Guinea pig
Monodelphis domestica
Ophiophagus hannah
Pteropus alecto
Rabbit
Rhesus monkey
Salmo salar
More Information
Related Products
miRBase Alias:
Human: hsa-let-7f-1* (v17)
Mouse: mmu-let-7f* (v15)
Mouse: mmu-let-7f-1* (v17)
Mouse: mmu-let-7f* (v15)
Mouse: mmu-let-7f-1* (v17)

Genomic Map