Assay Details
Gene Family ID: | MIPF0000097, mir-338 |
Mature miRNA Sequence: | UCCAGCAUCAGUGAUUUUGUUGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-338-3p | MIMAT0000581 |
Bovine | bta-miR-338 | MIMAT0009292 |
Chicken | gga-miR-338-3p | MIMAT0037552 |
Chrysemys picta | cpi-miR-338a-3p | MIMAT0037865 |
Columba livia | cli-miR-338b-3p | MIMAT0038621 |
Cricetulus griseus | cgr-miR-338 | MIMAT0023918 |
View More Rows | ||
Dasypus novemcinctus | dno-miR-338-3p | MIMAT0047820 |
Dog | cfa-miR-338 | MIMAT0006739 |
Gorilla gorilla | ggo-miR-338 | MIMAT0024125 |
Guinea pig | cpo-miR-338-3p | MIMAT0047175 |
Monodelphis domestica | mdo-miR-338 | MIMAT0004136 |
Ophiophagus hannah | oha-miR-338-3p | MIMAT0036904 |
Pteropus alecto | pal-miR-338-3p | MIMAT0040036 |
Python bivittatus | pbv-miR-338-3p | MIMAT0039042 |
Stem-loop Details
Rat
Bovine
Chicken
Chrysemys picta
Columba livia
Cricetulus griseus
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Monodelphis domestica
Ophiophagus hannah
Pteropus alecto
Python bivittatus
