Assay Details
Gene Family ID: | MIPF0000097, mir-338 |
Mature miRNA Sequence: | UCCAGCAUCAGUGAUUUUGUUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-338-3p | MIMAT0000581 |
| Bovine | bta-miR-338 | MIMAT0009292 |
| Chicken | gga-miR-338-3p | MIMAT0037552 |
| Chrysemys picta | cpi-miR-338a-3p | MIMAT0037865 |
| Columba livia | cli-miR-338b-3p | MIMAT0038621 |
| Cricetulus griseus | cgr-miR-338 | MIMAT0023918 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-338-3p | MIMAT0047820 |
| Dog | cfa-miR-338 | MIMAT0006739 |
| Gorilla gorilla | ggo-miR-338 | MIMAT0024125 |
| Guinea pig | cpo-miR-338-3p | MIMAT0047175 |
| Monodelphis domestica | mdo-miR-338 | MIMAT0004136 |
| Ophiophagus hannah | oha-miR-338-3p | MIMAT0036904 |
| Pteropus alecto | pal-miR-338-3p | MIMAT0040036 |
| Python bivittatus | pbv-miR-338-3p | MIMAT0039042 |
Stem-loop Details
Rat
Bovine
Chicken
Chrysemys picta
Columba livia
Cricetulus griseus
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Monodelphis domestica
Ophiophagus hannah
Pteropus alecto
Python bivittatus

Genomic Map