Assay Details
Gene Family ID: | MIPF0000013, mir-25 |
Mature miRNA Sequence: | CAUUGCACUUGUCUCGGUCUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-25-3p | MIMAT0000081 |
| Mouse | mmu-miR-25-3p | MIMAT0000652 |
| Rat | rno-miR-25-3p | MIMAT0000795 |
| Artibeus jamaicensis | aja-miR-25 | MIMAT0031142 |
| Astatotilapia burtoni | abu-miR-25 | MIMAT0042067 |
| Bovine | bta-miR-25 | MIMAT0003853 |
| View More Rows | ||
| Callithrix jacchus | cja-miR-25 | MIMAT0049317 |
| Cricetulus griseus | cgr-miR-25-3p | MIMAT0023870 |
| Cyprinus carpio | ccr-miR-25 | MIMAT0026280 |
| Dasypus novemcinctus | dno-miR-25-3p | MIMAT0047537 |
| Daubentonia madagascariensis | dma-miR-25 | MIMAT0049284 |
| Dog | cfa-miR-25 | MIMAT0006697 |
| Fugu rubripes | fru-miR-25 | MIMAT0002914 |
| Gadus morhua | gmo-miR-25-3p | MIMAT0044240 |
| Goat | chi-miR-25-3p | MIMAT0036100 |
| Gorilla gorilla | ggo-miR-25 | MIMAT0002760 |
| Guinea pig | cpo-miR-25-3p | MIMAT0046884 |
| Horse | eca-miR-25 | MIMAT0013057 |
| Ictalurus punctatus | ipu-miR-25 | MIMAT0029484 |
| Lagothrix lagotricha | lla-miR-25 | MIMAT0002779 |
| Macaca nemestrina | mne-miR-25 | MIMAT0002782 |
| Metriaclima zebra | mze-miR-25 | MIMAT0042300 |
| Monodelphis domestica | mdo-miR-25 | MIMAT0004179 |
| Neolamprologus brichardi | nbr-miR-25 | MIMAT0042482 |
| Nomascus leucogenys | nle-miR-25 | MIMAT0049352 |
| Oreochromis niloticus | oni-miR-25 | MIMAT0042669 |
| Otolemur garnettii | oga-miR-25 | MIMAT0049701 |
| Pan paniscus | ppa-miR-25 | MIMAT0002765 |
| Papio hamadryas | pha-miR-25 | MIMAT0049553 |
| Pongo pygmaeus | ppy-miR-25 | MIMAT0002768 |
| Pteropus alecto | pal-miR-25-3p | MIMAT0039823 |
| Pundamilia nyererei | pny-miR-25 | MIMAT0042872 |
| Rabbit | ocu-miR-25-3p | MIMAT0048134 |
| Rhesus monkey | mml-miR-25 | MIMAT0002774 |
| Salmo salar | ssa-miR-25-3p | MIMAT0032561 |
| Tetraodon nigroviridis | tni-miR-25 | MIMAT0002915 |
| Tupaia chinensis | tch-miR-25-3p | MIMAT0036590 |
| Xenopus laevis | xla-miR-25-3p | MIMAT0046554 |
| Xenopus tropicalis | xtr-miR-25 | MIMAT0003674 |
| Zebrafish | dre-miR-25-3p | MIMAT0001793 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-25 |
| Stem-loop Accession # | MI0000082 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | Chr. 7 - 100093560 - 100093643 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000081
miRBase ID: hsa-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: Chr. 7 - 100093560 - 100093643 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-25 |
| Stem-loop Accession # | MI0000689 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | Chr. 5 - 138165321 - 138165404 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000652
miRBase ID: mmu-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: Chr. 5 - 138165321 - 138165404 [-] on Build GRCm38 |
| Stem-loop ID | rno-mir-25 |
| Stem-loop Accession # | MI0000856 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACACGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | Chr. 12 - 19307340 - 19307423 [-] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000795
miRBase ID: rno-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: Chr. 12 - 19307340 - 19307423 [-] on Build Rnor_6.0 |
| Stem-loop ID | aja-mir-25 |
| Stem-loop Accession # | MI0024364 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0031142
miRBase ID: aja-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | abu-mir-25 |
| Stem-loop Accession # | MI0033761 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCCGGGCAUCCCAGAGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042067
miRBase ID: abu-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | bta-mir-25 |
| Stem-loop Accession # | MI0005067 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCCGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003853
miRBase ID: bta-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | cja-mir-25 |
| Stem-loop Accession # | MI0040053 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049317
miRBase ID: cja-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | cgr-mir-25 |
| Stem-loop Accession # | MI0020470 |
| Stem-loop Sequence | CGGGACUGGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGAAGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCCCAACACUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023870
miRBase ID: cgr-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ccr-mir-25 |
| Stem-loop Accession # | MI0023381 |
| Stem-loop Sequence | UAGCUGGUGUUGAGAGGCGGAGACUUGGGCAGUUGCUGGCUUUCCCACAAGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCACAACUCAUCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026280
miRBase ID: ccr-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | dno-mir-25 |
| Stem-loop Accession # | MI0038923 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047537
miRBase ID: dno-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | dma-mir-25 |
| Stem-loop Accession # | MI0040015 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049284
miRBase ID: dma-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | cfa-mir-25 |
| Stem-loop Accession # | MI0008111 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCGCGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006697
miRBase ID: cfa-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | fru-mir-25 |
| Stem-loop Accession # | MI0003229 |
| Stem-loop Sequence | GCUGGUGUUGAGAGGCGGAGACUUGGGCAAUUGCCGGCCAUCACAGAGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002914
miRBase ID: fru-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | gmo-mir-25 |
| Stem-loop Accession # | MI0036151 |
| Stem-loop Sequence | AGGCGGAGACUCGGGCAAUUGCCAGCCUUCCCAGACGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044240
miRBase ID: gmo-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | chi-mir-25 |
| Stem-loop Accession # | MI0030719 |
| Stem-loop Sequence | CGGGACUGGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCCGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCCCCACACUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036100
miRBase ID: chi-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ggo-mir-25 |
| Stem-loop Accession # | MI0003061 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002760
miRBase ID: ggo-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | cpo-mir-25 |
| Stem-loop Accession # | MI0038579 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046884
miRBase ID: cpo-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | eca-mir-25 |
| Stem-loop Accession # | MI0012801 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013057
miRBase ID: eca-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ipu-mir-25 |
| Stem-loop Accession # | MI0024599 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAGCUGCCGUCAUUCCCAGAAGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029484
miRBase ID: ipu-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | lla-mir-25 |
| Stem-loop Accession # | MI0003080 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002779
miRBase ID: lla-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | mne-mir-25 |
| Stem-loop Accession # | MI0003083 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002782
miRBase ID: mne-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | mze-mir-25 |
| Stem-loop Accession # | MI0034059 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCCGGGCAUCCCAGAGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042300
miRBase ID: mze-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | mdo-mir-25 |
| Stem-loop Accession # | MI0005370 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGAACUCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCUGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0004179
miRBase ID: mdo-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | nbr-mir-25 |
| Stem-loop Accession # | MI0034306 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCCGGGCAUCCCAGAGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042482
miRBase ID: nbr-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | nle-mir-25 |
| Stem-loop Accession # | MI0040089 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049352
miRBase ID: nle-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | oni-mir-25 |
| Stem-loop Accession # | MI0034568 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCCGGGCAUCCCAGAGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042669
miRBase ID: oni-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | oga-mir-25 |
| Stem-loop Accession # | MI0040485 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049701
miRBase ID: oga-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ppa-mir-25 |
| Stem-loop Accession # | MI0003066 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002765
miRBase ID: ppa-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | pha-mir-25 |
| Stem-loop Accession # | MI0040321 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049553
miRBase ID: pha-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ppy-mir-25 |
| Stem-loop Accession # | MI0003069 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002768
miRBase ID: ppy-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | pal-mir-25 |
| Stem-loop Accession # | MI0032347 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039823
miRBase ID: pal-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | pny-mir-25 |
| Stem-loop Accession # | MI0034828 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCCGGGCAUCCCAGAGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042872
miRBase ID: pny-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ocu-mir-25 |
| Stem-loop Accession # | MI0039238 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048134
miRBase ID: ocu-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | mml-mir-25 |
| Stem-loop Accession # | MI0003075 |
| Stem-loop Sequence | GGCCAGUGUUGAGAGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002774
miRBase ID: mml-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ssa-mir-25-1 |
| Stem-loop Accession # | MI0026646 |
| Stem-loop Sequence | AGGCGGAGUCUUGGGCAAUUGCCGGCAAUCUUAAGAGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032561
miRBase ID: ssa-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ssa-mir-25-2 |
| Stem-loop Accession # | MI0026647 |
| Stem-loop Sequence | AGGCGGAGUCUUGGGCAAUUGCCAGCAAUCUUAAGGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032561
miRBase ID: ssa-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | ssa-mir-25-3 |
| Stem-loop Accession # | MI0026648 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCCAGCCAUUCCAGACGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032561
miRBase ID: ssa-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | tni-mir-25 |
| Stem-loop Accession # | MI0003230 |
| Stem-loop Sequence | GCUGGUGUUGAGAGGCGGAGACUUGGGCAAUUGCCGGCCAUCACAGAGGGCAUUGCACUUGUCUCGGUCUGACAGUGCCGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002915
miRBase ID: tni-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | tch-mir-25 |
| Stem-loop Accession # | MI0031256 |
| Stem-loop Sequence | AGGCGGAGACUUGGGCAAUUGCUGGACGCUGCCCUGGGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036590
miRBase ID: tch-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | xla-mir-25 |
| Stem-loop Accession # | MI0038308 |
| Stem-loop Sequence | AGGCAGAGACAGGAGCAAUUGCUGGUGUACCUUGGGAGCAUUGCACUUGUCUCGGUCUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046554
miRBase ID: xla-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | xtr-mir-25-1 |
| Stem-loop Accession # | MI0004914 |
| Stem-loop Sequence | GCUGGUGUUGACAGGCAGAGACAGGAGCAACUGCUGGUGUGCCUUGGUAGCAUUGCACUUGUCUCGGUCUGACAGUGUCGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003674
miRBase ID: xtr-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | xtr-mir-25-2 |
| Stem-loop Accession # | MI0004915 |
| Stem-loop Sequence | GCUGGUGUUGACAGGCAGAGACAGGAGCAACUGCUGGUGUGCCUUGGUAGCAUUGCACUUGUCUCGGUCUGACAGUGUCGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003674
miRBase ID: xtr-miR-25 Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
| Stem-loop ID | dre-mir-25 |
| Stem-loop Accession # | MI0001922 |
| Stem-loop Sequence | GCCGGCGCUGAGAGGCGGAGACUUGGGCAGCUGCCGUCAUUCCCAGAAGGCAUUGCACUUGUCUCGGUCUGACAGUGGCGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001793
miRBase ID: dre-miR-25-3p Mature miRNA Sequence: CAUUGCACUUGUCUCGGUCUGA Chromosome Location: NA |
More Information
Related Products
miRBase Alias:
Human: hsa-miR-25 (v17)
Mouse: mmu-miR-25 (v17)
Rat: rno-miR-25 (v18)
Danio rerio: dre-miR-25 (v20)
Mouse: mmu-miR-25 (v17)
Rat: rno-miR-25 (v18)
Danio rerio: dre-miR-25 (v20)

Genomic Map