Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. 12 - 19307195 - 19307195 [-] on 19307195 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-25-5p | MIMAT0004713 |
| Rat | rno-miR-25-3p | MIMAT0000795 |
| Rat | rno-miR-93-5p | MIMAT0000817 |
| Rat | rno-miR-93-3p | MIMAT0017109 |
Stem-loop Details
Rat

Genomic Map