Assay Details
Gene Family ID: | MIPF0000128, mir-450 |
Mature miRNA Sequence: | UUUUGCAGUAUGUUCCUGAAUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-450b-5p | MIMAT0003511 |
| Cricetulus griseus | cgr-miR-450b-5p | MIMAT0023962 |
Stem-loop Details
Mouse
Cricetulus griseus

Genomic Map