Assay Details
| Gene Family ID: | MIPF0000128, mir-450 | 
| Mature miRNA Sequence: | UUUUGCAGUAUGUUCCUGAAUA | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-450b-5p | MIMAT0003511 | 
| Cricetulus griseus | cgr-miR-450b-5p | MIMAT0023962 | 
Stem-loop Details
Mouse
Cricetulus griseus
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :