Assay Details
Gene Family ID: | MIPF0000099, mir-136 |
Mature miRNA Sequence: | ACUCCAUUUGUUUUGAUGAUGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-136-5p | MIMAT0000448 |
| Rat | rno-miR-136-5p | MIMAT0000842 |
| Bovine | bta-miR-136 | MIMAT0009230 |
| Dasypus novemcinctus | dno-miR-136-5p | MIMAT0047644 |
| Dog | cfa-miR-136 | MIMAT0006714 |
| Gorilla gorilla | ggo-miR-136 | MIMAT0002293 |
| View More Rows | ||
| Guinea pig | cpo-miR-136-5p | MIMAT0046997 |
| Pan paniscus | ppa-miR-136 | MIMAT0002295 |
| Pan troglodytes | ptr-miR-136 | MIMAT0002292 |
| Pig | ssc-miR-136-5p | MIMAT0002158 |
| Pongo pygmaeus | ppy-miR-136 | MIMAT0002294 |
| Pteropus alecto | pal-miR-136-5p | MIMAT0040001 |
| Rabbit | ocu-miR-136-5p | MIMAT0048232 |
| Rhesus monkey | mml-miR-136 | MIMAT0006192 |
| Sheep | oar-miR-136 | MIMAT0001417 |
| Tupaia chinensis | tch-miR-136-5p | MIMAT0036531 |
Stem-loop Details
Human
Rat
Bovine
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Sheep
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-136 (v17)
Rat: rno-miR-136 (v18)
Staphylococcus schleiferi: ssc-miR-136 (v21)
Rat: rno-miR-136 (v18)
Staphylococcus schleiferi: ssc-miR-136 (v21)

Genomic Map