Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 6 - 133717111 - 133717111 [-] on 133717111 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-136-5p | MIMAT0000842 |
| Rat | rno-miR-136-3p | MIMAT0004733 |
Stem-loop Details
Rat

Genomic Map