Assay Details
Gene Family ID: | MIPF0000185, mir-542 |
Mature miRNA Sequence: | CUCGGGGAUCAUCAUGUCACGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-542-5p | MIMAT0003171 |
Rat | rno-miR-542-5p | MIMAT0003178 |
Cricetulus griseus | cgr-miR-542-5p | MIMAT0023983 |
Horse | eca-miR-542-5p | MIMAT0013237 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Horse
