Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. X - 158588762 - 158588762 [-] on 158588762 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-542-5p | MIMAT0003178 |
| Rat | rno-miR-542-3p | MIMAT0003179 |
Stem-loop Details
Rat

Genomic Map