Assay Details
Amplicon Length: | 97 |
Chromosome Location: | Chr. X - 53049696 - 53049696 [-] on 53049696 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-542-5p | MIMAT0003171 |
| Mouse | mmu-miR-542-3p | MIMAT0003172 |
Stem-loop Details
Mouse

Genomic Map