Assay Details
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | AAGACGGGAGAAGAGAAGGGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-483-5p | MIMAT0004782 |
| Rat | rno-miR-483-5p | MIMAT0017201 |
| Guinea pig | cpo-miR-483-5p | MIMAT0047273 |
Stem-loop Details
Mouse
Rat
Guinea pig
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-483 (v17)
Rat: rno-miR-483* (v18)
Rat: rno-miR-483* (v18)

Genomic Map