Assay Details
Gene Family ID: | MIPF0000164, mir-322 |
Mature miRNA Sequence: | CAAAACGUGAGGCGCUGCUAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-424-3p | MIMAT0004749 |
| Bovine | bta-miR-424-3p | MIMAT0015304 |
| Dog | cfa-miR-424 | MIMAT0006745 |
| Goat | chi-miR-424-3p | MIMAT0036223 |
| Pig | ssc-miR-424-3p | MIMAT0013921 |
| Pteropus alecto | pal-miR-424-3p | MIMAT0039920 |
Stem-loop Details
Human
Bovine
Dog
Goat
Pig
Pteropus alecto
More Information
Related Products
miRBase Alias:
Human: hsa-miR-424* (v17)
Bos taurus: bta-miR-424* (v18)
Staphylococcus schleiferi: ssc-miR-424* (v18)
Bos taurus: bta-miR-424* (v18)
Staphylococcus schleiferi: ssc-miR-424* (v18)

Genomic Map