Assay Details
Gene Family ID: | MIPF0000164, mir-322 | 
Mature miRNA Sequence: | CAAAACGUGAGGCGCUGCUAU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-424-3p | MIMAT0004749 | 
| Bovine | bta-miR-424-3p | MIMAT0015304 | 
| Dog | cfa-miR-424 | MIMAT0006745 | 
| Goat | chi-miR-424-3p | MIMAT0036223 | 
| Pig | ssc-miR-424-3p | MIMAT0013921 | 
| Pteropus alecto | pal-miR-424-3p | MIMAT0039920 | 
Stem-loop Details
Human
Bovine
Dog
Goat
Pig
Pteropus alecto
More Information
Related Products
miRBase Alias:
						
							Human: hsa-miR-424* (v17)
Bos taurus: bta-miR-424* (v18)
Staphylococcus schleiferi: ssc-miR-424* (v18)
						
					
				Bos taurus: bta-miR-424* (v18)
Staphylococcus schleiferi: ssc-miR-424* (v18)

Genomic Map