Assay Details
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UAGGUAGUUUCGUGUUGUUGGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-196c-5p | MIMAT0005303 |
| Cricetulus griseus | cgr-miR-196c | MIMAT0023834 |
Stem-loop Details
Rat
Cricetulus griseus

Genomic Map