Assay Details
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UAGGUAGUUUCGUGUUGUUGGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-196c-5p | MIMAT0005303 |
Cricetulus griseus | cgr-miR-196c | MIMAT0023834 |
Stem-loop Details
Rat
Cricetulus griseus
