Assay Details
Amplicon Length: | 57 |
Chromosome Location: | Chr. 10 - 84113332 - 84113332 [-] on 84113332 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-196c-3p | MIMAT0017299 |
Rat | rno-miR-196c-5p | MIMAT0005303 |
Stem-loop Details
Rat
