Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UUGAAGAGAGGUUAUCCUUUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-300-5p | MIMAT0004578 |
| Rat | rno-miR-300-5p | MIMAT0004743 |
Stem-loop Details
Mouse
Rat

Genomic Map