Assay Details
Amplicon Length: | 80 |
Chromosome Location: | Chr. 12 - 109593147 - 109593147 [-] on 109593147 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-127-5p | MIMAT0004530 |
| Mouse | mmu-miR-127-3p | MIMAT0000139 |
Stem-loop Details
Mouse

Genomic Map