Assay Details
Gene Family ID: | MIPF0000080, mir-127 |
Mature miRNA Sequence: | UCGGAUCCGUCUGAGCUUGGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-127-3p | MIMAT0000446 |
| Mouse | mmu-miR-127-3p | MIMAT0000139 |
| Rat | rno-miR-127-3p | MIMAT0000833 |
| Ateles geoffroyi | age-miR-127 | MIMAT0002287 |
| Bovine | bta-miR-127 | MIMAT0003787 |
| Cricetulus griseus | cgr-miR-127 | MIMAT0041149 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-127-3p | MIMAT0047620 |
| Daubentonia madagascariensis | dma-miR-127 | MIMAT0049248 |
| Dog | cfa-miR-127 | MIMAT0006713 |
| Guinea pig | cpo-miR-127-3p | MIMAT0046973 |
| Horse | eca-miR-127 | MIMAT0013126 |
| Lagothrix lagotricha | lla-miR-127 | MIMAT0002285 |
| Macaca nemestrina | mne-miR-127 | MIMAT0002286 |
| Nomascus leucogenys | nle-miR-127 | MIMAT0049373 |
| Otolemur garnettii | oga-miR-127 | MIMAT0049678 |
| Pan paniscus | ppa-miR-127 | MIMAT0049050 |
| Pan troglodytes | ptr-miR-127 | MIMAT0002282 |
| Papio hamadryas | pha-miR-127 | MIMAT0049580 |
| Pig | ssc-miR-127 | MIMAT0013932 |
| Pongo pygmaeus | ppy-miR-127 | MIMAT0002283 |
| Pteropus alecto | pal-miR-127-3p | MIMAT0040147 |
| Rabbit | ocu-miR-127-3p | MIMAT0048209 |
| Rhesus monkey | mml-miR-127-3p | MIMAT0002281 |
| Saguinus labiatus | sla-miR-127 | MIMAT0002284 |
| Saimiri boliviensis | sbo-miR-127 | MIMAT0049479 |
| Tupaia chinensis | tch-miR-127-3p | MIMAT0036559 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-127 |
| Stem-loop Accession # | MI0000472 |
| Stem-loop Sequence | UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC |
| Chromosome Location | Chr. 14 - 100882979 - 100883075 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000446
miRBase ID: hsa-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: Chr. 14 - 100882979 - 100883075 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-127 |
| Stem-loop Accession # | MI0000154 |
| Stem-loop Sequence | CCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGG |
| Chromosome Location | Chr. 12 - 109592846 - 109592915 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000139
miRBase ID: mmu-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: Chr. 12 - 109592846 - 109592915 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-127 |
| Stem-loop Accession # | MI0000899 |
| Stem-loop Sequence | UUUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC |
| Chromosome Location | Chr. 6 - 133713430 - 133713526 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000833
miRBase ID: rno-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: Chr. 6 - 133713430 - 133713526 [+] on Build Rnor_6.0 |
| Stem-loop ID | age-mir-127 |
| Stem-loop Accession # | MI0002588 |
| Stem-loop Sequence | UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002287
miRBase ID: age-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | bta-mir-127 |
| Stem-loop Accession # | MI0005008 |
| Stem-loop Sequence | UGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCCUCAUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003787
miRBase ID: bta-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | cgr-mir-127 |
| Stem-loop Accession # | MI0033078 |
| Stem-loop Sequence | CCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0041149
miRBase ID: cgr-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | dno-mir-127 |
| Stem-loop Accession # | MI0038969 |
| Stem-loop Sequence | CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047620
miRBase ID: dno-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | dma-mir-127 |
| Stem-loop Accession # | MI0039976 |
| Stem-loop Sequence | CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049248
miRBase ID: dma-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | cfa-mir-127 |
| Stem-loop Accession # | MI0008132 |
| Stem-loop Sequence | CCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006713
miRBase ID: cfa-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-127 |
| Stem-loop Accession # | MI0038628 |
| Stem-loop Sequence | CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046973
miRBase ID: cpo-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | eca-mir-127 |
| Stem-loop Accession # | MI0012876 |
| Stem-loop Sequence | CCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013126
miRBase ID: eca-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | lla-mir-127 |
| Stem-loop Accession # | MI0002586 |
| Stem-loop Sequence | UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002285
miRBase ID: lla-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | mne-mir-127 |
| Stem-loop Accession # | MI0002587 |
| Stem-loop Sequence | UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCCUCAUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002286
miRBase ID: mne-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | nle-mir-127 |
| Stem-loop Accession # | MI0040110 |
| Stem-loop Sequence | CCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049373
miRBase ID: nle-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | oga-mir-127 |
| Stem-loop Accession # | MI0040461 |
| Stem-loop Sequence | CCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049678
miRBase ID: oga-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | ppa-mir-127 |
| Stem-loop Accession # | MI0039758 |
| Stem-loop Sequence | CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049050
miRBase ID: ppa-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | ptr-mir-127 |
| Stem-loop Accession # | MI0002583 |
| Stem-loop Sequence | UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002282
miRBase ID: ptr-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | pha-mir-127 |
| Stem-loop Accession # | MI0040355 |
| Stem-loop Sequence | UGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049580
miRBase ID: pha-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | ssc-mir-127 |
| Stem-loop Accession # | MI0013144 |
| Stem-loop Sequence | AUCGCUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013932
miRBase ID: ssc-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | ppy-mir-127 |
| Stem-loop Accession # | MI0002584 |
| Stem-loop Sequence | UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002283
miRBase ID: ppy-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | pal-mir-127 |
| Stem-loop Accession # | MI0032517 |
| Stem-loop Sequence | CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0040147
miRBase ID: pal-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | ocu-mir-127 |
| Stem-loop Accession # | MI0039280 |
| Stem-loop Sequence | CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048209
miRBase ID: ocu-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | mml-mir-127 |
| Stem-loop Accession # | MI0002582 |
| Stem-loop Sequence | UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCCUCAUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002281
miRBase ID: mml-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | sla-mir-127 |
| Stem-loop Accession # | MI0002585 |
| Stem-loop Sequence | UGUGAUCACUGUCUCCAGCCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCUGGUCGGAAGUCUCAUCAUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002284
miRBase ID: sla-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | sbo-mir-127 |
| Stem-loop Accession # | MI0040236 |
| Stem-loop Sequence | CCUGCUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049479
miRBase ID: sbo-miR-127 Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
| Stem-loop ID | tch-mir-127 |
| Stem-loop Accession # | MI0031219 |
| Stem-loop Sequence | CUGAAGCUCAGAGGGCUCUGAUUCAGAAAGAUCAUCGGAUCCGUCUGAGCUUGGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036559
miRBase ID: tch-miR-127-3p Mature miRNA Sequence: UCGGAUCCGUCUGAGCUUGGCU Chromosome Location: NA |
More Information
Related Products

Genomic Map