Assay Details
Gene Family ID: | MIPF0000080, mir-127 |
Mature miRNA Sequence: | UCGGAUCCGUCUGAGCUUGGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-127-3p | MIMAT0000446 |
| Mouse | mmu-miR-127-3p | MIMAT0000139 |
| Rat | rno-miR-127-3p | MIMAT0000833 |
| Ateles geoffroyi | age-miR-127 | MIMAT0002287 |
| Bovine | bta-miR-127 | MIMAT0003787 |
| Cricetulus griseus | cgr-miR-127 | MIMAT0041149 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-127-3p | MIMAT0047620 |
| Daubentonia madagascariensis | dma-miR-127 | MIMAT0049248 |
| Dog | cfa-miR-127 | MIMAT0006713 |
| Guinea pig | cpo-miR-127-3p | MIMAT0046973 |
| Horse | eca-miR-127 | MIMAT0013126 |
| Lagothrix lagotricha | lla-miR-127 | MIMAT0002285 |
| Macaca nemestrina | mne-miR-127 | MIMAT0002286 |
| Nomascus leucogenys | nle-miR-127 | MIMAT0049373 |
| Otolemur garnettii | oga-miR-127 | MIMAT0049678 |
| Pan paniscus | ppa-miR-127 | MIMAT0049050 |
| Pan troglodytes | ptr-miR-127 | MIMAT0002282 |
| Papio hamadryas | pha-miR-127 | MIMAT0049580 |
| Pig | ssc-miR-127 | MIMAT0013932 |
| Pongo pygmaeus | ppy-miR-127 | MIMAT0002283 |
| Pteropus alecto | pal-miR-127-3p | MIMAT0040147 |
| Rabbit | ocu-miR-127-3p | MIMAT0048209 |
| Rhesus monkey | mml-miR-127-3p | MIMAT0002281 |
| Saguinus labiatus | sla-miR-127 | MIMAT0002284 |
| Saimiri boliviensis | sbo-miR-127 | MIMAT0049479 |
| Tupaia chinensis | tch-miR-127-3p | MIMAT0036559 |
Stem-loop Details
Human
Mouse
Rat
Ateles geoffroyi
Bovine
Cricetulus griseus
Dasypus novemcinctus
Daubentonia madagascariensis
Dog
Guinea pig
Horse
Lagothrix lagotricha
Macaca nemestrina
Nomascus leucogenys
Otolemur garnettii
Pan paniscus
Pan troglodytes
Papio hamadryas
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Saguinus labiatus
Saimiri boliviensis
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-127 (v9.2)
Mouse: mmu-miR-127 (v17)
Rat: rno-miR-127 (v18)
Macaca mulatta: mml-miR-127 (v19)
Mouse: mmu-miR-127 (v17)
Rat: rno-miR-127 (v18)
Macaca mulatta: mml-miR-127 (v19)

Genomic Map