Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 13 - 48537430 - 48537430 [+] on 48537430 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-let-7f-1-3p | MIMAT0004623 |
| Mouse | mmu-let-7f-5p | MIMAT0000525 |
Stem-loop Details
Mouse

Genomic Map