Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 7 - 61877563 - 61877563 [+] on 61877563 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-344-5p | MIMAT0017038 |
| Mouse | mmu-miR-344-3p | MIMAT0000593 |
Stem-loop Details
Mouse

Genomic Map