Assay Details
Gene Family ID: | MIPF0000267, mir-344 | 
Mature miRNA Sequence: | UGAUCUAGCCAAAGCCUGACUGU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-344-3p | MIMAT0000593 | 
| Rat | rno-miR-344a-3p | MIMAT0000592 | 
| Cricetulus griseus | cgr-miR-344 | MIMAT0023924 | 
Stem-loop Details
Mouse
Rat
Cricetulus griseus

Genomic Map