Assay Details
Gene Family ID: | MIPF0000267, mir-344 |
Mature miRNA Sequence: | UGAUCUAGCCAAAGCCUGACUGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-344-3p | MIMAT0000593 |
Rat | rno-miR-344a-3p | MIMAT0000592 |
Cricetulus griseus | cgr-miR-344 | MIMAT0023924 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
