Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | AUAAGACGAGCAAAAAGCUUGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-208a-3p | MIMAT0000241 |
Mouse | mmu-miR-208a-3p | MIMAT0000520 |
Anolis carolinensis | aca-miR-208-3p | MIMAT0021860 |
Bovine | bta-miR-208a | MIMAT0009261 |
Callithrix jacchus | cja-miR-208a | MIMAT0039597 |
Dasypus novemcinctus | dno-miR-208a-3p | MIMAT0047751 |
View More Rows | ||
Dog | cfa-miR-208a | MIMAT0009868 |
Guinea pig | cpo-miR-208a-3p | MIMAT0047106 |
Horse | eca-miR-208a | MIMAT0012899 |
Ophiophagus hannah | oha-miR-208-3p | MIMAT0036821 |
Ornithorhynchus anatinus | oan-miR-208-3p | MIMAT0006921 |
Pan troglodytes | ptr-miR-208a | MIMAT0008067 |
Pongo pygmaeus | ppy-miR-208a | MIMAT0015799 |
Pteropus alecto | pal-miR-208a-3p | MIMAT0040026 |
Rabbit | ocu-miR-208a-3p | MIMAT0048330 |
Rhesus monkey | mml-miR-208a-3p | MIMAT0006238 |
Stem-loop Details
Human
Mouse
Anolis carolinensis
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Ophiophagus hannah
Ornithorhynchus anatinus
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-208a (v19)
Human: hsa-miR-208 (v10)
Mouse: mmu-miR-208 (v10)
Mouse: mmu-miR-208a (v15)
Rat: rno-miR-208 (v15)
Aeromonas caviae Ae398: aca-miR-208 (v18)
Macaca mulatta: mml-miR-208a (v19)
Ochrobactrum anthropi: oan-miR-208 (v18)
Human: hsa-miR-208 (v10)
Mouse: mmu-miR-208 (v10)
Mouse: mmu-miR-208a (v15)
Rat: rno-miR-208 (v15)
Aeromonas caviae Ae398: aca-miR-208 (v18)
Macaca mulatta: mml-miR-208a (v19)
Ochrobactrum anthropi: oan-miR-208 (v18)
