Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 14 - 54948863 - 54948863 [+] on 54948863 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-208a-3p | MIMAT0000520 |
Mouse | mmu-miR-208a-5p | MIMAT0017014 |
Stem-loop Details
Mouse
