Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | AUAAGACGAGCAAAAAGCUUGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-208a-3p | MIMAT0000241 |
Mouse | mmu-miR-208a-3p | MIMAT0000520 |
Anolis carolinensis | aca-miR-208-3p | MIMAT0021860 |
Bovine | bta-miR-208a | MIMAT0009261 |
Callithrix jacchus | cja-miR-208a | MIMAT0039597 |
Dasypus novemcinctus | dno-miR-208a-3p | MIMAT0047751 |
View More Rows | ||
Dog | cfa-miR-208a | MIMAT0009868 |
Guinea pig | cpo-miR-208a-3p | MIMAT0047106 |
Horse | eca-miR-208a | MIMAT0012899 |
Ophiophagus hannah | oha-miR-208-3p | MIMAT0036821 |
Ornithorhynchus anatinus | oan-miR-208-3p | MIMAT0006921 |
Pan troglodytes | ptr-miR-208a | MIMAT0008067 |
Pongo pygmaeus | ppy-miR-208a | MIMAT0015799 |
Pteropus alecto | pal-miR-208a-3p | MIMAT0040026 |
Rabbit | ocu-miR-208a-3p | MIMAT0048330 |
Rhesus monkey | mml-miR-208a-3p | MIMAT0006238 |
Stem-loop Details
Human
Mouse
Anolis carolinensis
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Ophiophagus hannah
Ornithorhynchus anatinus
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
