Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 6 - 133862516 - 133862516 [+] on 133862516 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-329-5p | MIMAT0017031 |
| Rat | rno-miR-329-3p | MIMAT0000566 |
Stem-loop Details
Rat

Genomic Map