Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AACACACCCAGCUAACCUUUUU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-329-3p | MIMAT0000567 |
Rat | rno-miR-329-3p | MIMAT0000566 |
Stem-loop Details
Mouse
Rat

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-329 (v17)
Rat: rno-miR-329 (v18)
Rat: rno-miR-329 (v18)
