Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AACACACCCAGCUAACCUUUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-329-3p | MIMAT0000567 |
| Rat | rno-miR-329-3p | MIMAT0000566 |
Stem-loop Details
Mouse
Rat

Genomic Map