Assay Details
Gene Family ID: | MIPF0000199, mir-331 |
Mature miRNA Sequence: | GCCCCUGGGCCUAUCCUAGAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-331-3p | MIMAT0000760 |
| Mouse | mmu-miR-331-3p | MIMAT0000571 |
| Rat | rno-miR-331-3p | MIMAT0000570 |
| Artibeus jamaicensis | aja-miR-331 | MIMAT0031150 |
| Bovine | bta-miR-331-3p | MIMAT0004339 |
| Dasypus novemcinctus | dno-miR-331-3p | MIMAT0047814 |
| View More Rows | ||
| Dog | cfa-miR-331 | MIMAT0009895 |
| Guinea pig | cpo-miR-331-3p | MIMAT0047169 |
| Horse | eca-miR-331 | MIMAT0013188 |
| Pan troglodytes | ptr-miR-331 | MIMAT0008103 |
| Pig | ssc-miR-331-3p | MIMAT0013930 |
| Pteropus alecto | pal-miR-331-3p | MIMAT0039994 |
| Rabbit | ocu-miR-331-3p | MIMAT0048381 |
| Rhesus monkey | mml-miR-331-3p | MIMAT0006273 |
Stem-loop Details
Human
Mouse
Rat
Artibeus jamaicensis
Bovine
Dasypus novemcinctus
Dog
Guinea pig
Horse
Pan troglodytes
Pig
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map